3-(2-Chloro-phenyl)-1H-pyrrole 90%
Dodavatel: Apollo Scientific
OR480256-250MG
OR480256-500MG
APOSOR480256-250MGEA
21560
CZK
APOSOR480256-250MG
APOSOR480256-500MG
3-(2-Chloro-phenyl)-1H-pyrrole 90%
3-(2-Chloro-phenyl)-1H-pyrrole
Vzorec:
C₁₀H₈ClN MW: 177,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832514 CAS číslo: 71845-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...