4-(1-Methylpyrazol-4-yl)aniline
Dodavatel: Apollo Scientific
Synonyms:
4-(1-Methyl-1H-pyrazol-4-yl)aniline
OR480242-1G
OR480242-250MG
OR480242-500MG
APOSOR480242-1GEA
10620
CZK
APOSOR480242-1G
APOSOR480242-250MG
APOSOR480242-500MG
4-(1-Methylpyrazol-4-yl)aniline
4-(1-Methylpyrazol-4-yl)aniline
Vzorec:
C₁₀H₁₁N₃ MW: 173,214 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797404 CAS číslo: 1079178-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...