Tisk…

2-Methyloxazole-5-carbaldehyde ≥97%

Dodavatel: Apollo Scientific

OR480235-1G OR480235-250MG
APOSOR480235-1GEA 10820 CZK
APOSOR480235-1G APOSOR480235-250MG
2-Methyloxazole-5-carbaldehyde ≥97%
2-Methyloxazole-5-carbaldehyde
Vzorec: C₅H₅NO₂
MW: 111,099 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738689
CAS číslo: 885273-42-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR