Tisk…

1,3,4-Oxadiazol-2-ylmethanamine hydrochloride

Dodavatel: Apollo Scientific

OR480234-1G OR480234-250MG OR480234-500MG OR480234-100MG
APOSOR480234-1GEA 0 CZK
APOSOR480234-1G APOSOR480234-250MG APOSOR480234-500MG APOSOR480234-100MG
1,3,4-Oxadiazol-2-ylmethanamine hydrochloride
1,3,4-Oxadiazol-2-ylmethanamine hydrochloride
Vzorec: C₃H₅N₃O·ClH
Storage Temperature: Ambient
MDL Number: MFCD13185270
CAS číslo: 1383788-35-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR