2-Cyano-3,6-dimethylpyridine 95%
Dodavatel: Apollo Scientific
OR480233-1G
OR480233-250MG
OR480233-5G
APOSOR480233-1GEA
16240
CZK
APOSOR480233-1G
APOSOR480233-250MG
APOSOR480233-5G
2-Cyano-3,6-dimethylpyridine 95%
2-Cyano-3,6-dimethylpyridine
Vzorec:
C₈H₈N₂ MW: 132,163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06637568 CAS číslo: 68164-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...