6-Pyrrolidin-1-ylpyridine-2-carbaldehyde 97%
Dodavatel: Apollo Scientific
OR47916-10G
OR47916-1G
OR47916-250MG
APOSOR47916-10GEA
13890
CZK
APOSOR47916-10G
APOSOR47916-1G
APOSOR47916-250MG
6-Pyrrolidin-1-ylpyridine-2-carbaldehyde 97%
6-Pyrrolidin-1-ylpyridine-2-carbaldehyde
Vzorec:
C₁₀H₁₂N₂O MW: 176,215 g/mol Bod tání: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD08435859 CAS číslo: 230618-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...