2-Bromo-5-chloro-4-(hydroxymethyl)pyridine 97%
Dodavatel: Apollo Scientific
OR47898-1G
OR47898-5G
APOSOR47898-1GEA
9100
CZK
APOSOR47898-1G
APOSOR47898-5G
2-Bromo-5-chloro-4-(hydroxymethyl)pyridine 97%
2-Bromo-5-chloro-4-(hydroxymethyl)pyridine
Vzorec:
C₆H₅BrClNO Storage Temperature: Ambient |
MDL Number:
MFCD16606985 CAS číslo: 1227599-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...