Tisk…

Diethyl 2-(3-cyanopropyl)malonate

Dodavatel: Apollo Scientific

OR47892-1G OR47892-5G
APOSOR47892-1GEA 622 CZK
APOSOR47892-1G APOSOR47892-5G
Diethyl 2-(3-cyanopropyl)malonate
Diethyl 2-(3-cyanopropyl)malonate
Vzorec: C₁₁H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD07357261
CAS číslo: 63972-18-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR