3-Amino-4-cyanobenzoic acid 97%
Dodavatel: Apollo Scientific
OR47858-1G
APOSOR47858-1GEA
2940
CZK
APOSOR47858-1G
3-Amino-4-cyanobenzoic acid 97%
3-Amino-4-cyanobenzoic acid
Vzorec:
C₈H₆N₂O₂ MW: 162,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20640420 CAS číslo: 159847-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...