Tisk…

trans-4-(Morpholin-4-yl)cyclohexylamine dihydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: trans-4-Morpholinocyclohexanamine dihydrochloride, (1r,4r)-4-Morpholinocyclohexanamine dihydrochloride

OR47855-100MG OR47855-1G OR47855-250MG
APOSOR47855-100MGEA 2900 CZK
APOSOR47855-100MG APOSOR47855-1G APOSOR47855-250MG
trans-4-(Morpholin-4-yl)cyclohexylamine dihydrochloride 98%
trans-4-(Morpholin-4-yl)cyclohexylamine dihydrochloride
Vzorec: C₁₀H₂₀N₂O·2ClH
Storage Temperature: Ambient
MDL Number: MFCD27978481
CAS číslo: 412356-24-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR