2-Bromo-5-cyclopropylnitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
OR47842-1G
OR47842-250MG
OR47842-5G
APOSOR47842-1GEA
14880
CZK
APOSOR47842-1G
APOSOR47842-250MG
APOSOR47842-5G
2-Bromo-5-cyclopropylnitrobenzene
2-Bromo-5-cyclopropylnitrobenzene
Vzorec:
C₉H₈BrNO₂ MW: 242.069 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD32264080 CAS číslo: 2387348-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...