(4-Chloro-1-cyclopropyl-1H-pyrazol-3-yl)methanol
Dodavatel: Apollo Scientific
OR475190-1G
OR475190-250MG
APOSOR475190-1GEA
18620
CZK
APOSOR475190-1G
APOSOR475190-250MG
(4-Chloro-1-cyclopropyl-1H-pyrazol-3-yl)methanol
(4-Chloro-1-cyclopropyl-1H-pyrazol-3-yl)methanol
Vzorec:
C₇H₉ClN₂O MW: 172,612 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD32200422 CAS číslo: 2413937-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...