1-Cyclopropyl-1H-pyrazole-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR475183-1G
OR475183-250MG
OR475183-100MG
APOSOR475183-1GEA
17360
CZK
APOSOR475183-1G
APOSOR475183-250MG
APOSOR475183-100MG
1-Cyclopropyl-1H-pyrazole-3-carboxylic acid
1-Cyclopropyl-1H-pyrazole-3-carboxylic acid
Vzorec:
C₇H₈N₂O₂ MW: 152,151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26520322 CAS číslo: 1403333-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...