Tisk…

[1-(4-Bromophenyl)-1H-pyrazol-3-yl]methanol

Dodavatel: Apollo Scientific
Varování

OR475179-100MG OR475179-1G OR475179-250MG
APOSOR475179-100MGEA 744 CZK
APOSOR475179-100MG APOSOR475179-1G APOSOR475179-250MG
[1-(4-Bromophenyl)-1H-pyrazol-3-yl]methanol
[1-(4-Bromophenyl)-1H-pyrazol-3-yl]methanol
Vzorec: C₁₀H₉BrN₂O
MW: 253,095 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21304384
CAS číslo: 1478538-20-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR