Tisk…

Thieno[2,3-c]pyridine-4-boronic acid, pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[2,3-c]pyridine

OR475157-100MG OR475157-250MG
APOSOR475157-100MGEA 12200 CZK
APOSOR475157-100MG APOSOR475157-250MG
Thieno[2,3-c]pyridine-4-boronic acid, pinacol ester
Thieno[2,3-c]pyridine-4-boronic acid, pinacol ester
Vzorec: C₁₃H₁₆BNO₂S
MW: 261.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26517928
CAS číslo: 2230209-53-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR