Tisk…

(1R)-1-(3-Bromophenyl)-N-methyl-ethanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR475155-1G OR475155-250MG
APOSOR475155-1GEA 16370 CZK
APOSOR475155-1G APOSOR475155-250MG
(1R)-1-(3-Bromophenyl)-N-methyl-ethanamine hydrochloride
(1R)-1-(3-Bromophenyl)-N-methyl-ethanamine hydrochloride
Vzorec: C₉H₁₂BrN·ClH
MW: 250,563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31714839
CAS číslo: 2366997-08-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR