Tisk…

1-(2-Bromophenyl)-N-methyl-ethanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR475085-100MG OR475085-1G OR475085-250MG
APOSOR475085-100MGEA 6780 CZK
APOSOR475085-100MG APOSOR475085-1G APOSOR475085-250MG
1-(2-Bromophenyl)-N-methyl-ethanamine hydrochloride
1-(2-Bromophenyl)-N-methyl-ethanamine hydrochloride
Vzorec: C₉H₁₂BrN·ClH
MW: 250,563 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22566065
CAS číslo: 1423025-56-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR