Tisk…

(1S)-1-(4-Bromophenyl)-N-methylethanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-1-(4-Bromophenyl)-N-methylethanamine hydrochloride

OR475072-100MG OR475072-250MG
APOSOR475072-100MGEA 4400 CZK
APOSOR475072-100MG APOSOR475072-250MG
(1S)-1-(4-Bromophenyl)-N-methylethanamine hydrochloride
(1S)-1-(4-Bromophenyl)-N-methylethanamine hydrochloride
Vzorec: C₉H₁₂BrN·ClH
MW: 250.563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977502
CAS číslo: 1810074-73-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR