Tisk…

4-Bromo-3-chloro-2-methylpyridine 95%

Dodavatel: Apollo Scientific
Varování

OR471724-100MG OR471724-1G OR471724-250MG
APOSOR471724-100MGEA 4070 CZK
APOSOR471724-100MG APOSOR471724-1G APOSOR471724-250MG
4-Bromo-3-chloro-2-methylpyridine 95%
4-Bromo-3-chloro-2-methylpyridine
Vzorec: C₆H₅BrClN
MW: 206,468 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11215565
CAS číslo: 1188140-52-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR