2-(4-Bromophenyl)tetrahydrofuran 95%
Dodavatel: Apollo Scientific
OR471716-1G
OR471716-250MG
APOSOR471716-1GEA
37690
CZK
APOSOR471716-1G
APOSOR471716-250MG
2-(4-Bromophenyl)tetrahydrofuran 95%
2-(4-Bromophenyl)tetrahydrofuran
Vzorec:
C₁₀H₁₁BrO Storage Temperature: Ambient |
MDL Number:
MFCD19320036 CAS číslo: 194725-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...