Tisk…

4'-Amino-3'-chloroacetophenone 95%

Dodavatel: Apollo Scientific

OR471714-1G OR471714-250MG
APOSOR471714-1GEA 22820 CZK
APOSOR471714-1G APOSOR471714-250MG
4'-Amino-3'-chloroacetophenone 95%
4'-Amino-3'-chloroacetophenone
Vzorec: C₈H₈ClNO
MW: 169,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411008
CAS číslo: 6953-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR