1H-Pyrrolo[2,3-b]pyridine-4-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR471713-1G
OR471713-250MG
APOSOR471713-1GEA
5060
CZK
APOSOR471713-1G
APOSOR471713-250MG
1H-Pyrrolo[2,3-b]pyridine-4-carbaldehyde 95%
1H-Pyrrolo[2,3-b]pyridine-4-carbaldehyde
Vzorec:
C₈H₆N₂O MW: 146,146 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08272237 CAS číslo: 728034-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...