Tisk…

3-Hydroxy-6-methylpyridine-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR471706-1G OR471706-250MG
APOSOR471706-1GEA 22820 CZK
APOSOR471706-1G APOSOR471706-250MG
3-Hydroxy-6-methylpyridine-2-carbaldehyde 95%
3-Hydroxy-6-methylpyridine-2-carbaldehyde
Vzorec: C₇H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD11044356
CAS číslo: 66497-42-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR