Tisk…

4'-Amino-3'-iodoacetophenone 97%

Dodavatel: Apollo Scientific

OR471702-1G OR471702-5G
APOSOR471702-1GEA 10490 CZK
APOSOR471702-1G APOSOR471702-5G
4'-Amino-3'-iodoacetophenone 97%
4'-Amino-3'-iodoacetophenone
Vzorec: C₈H₈INO
Storage Temperature: Ambient
MDL Number: MFCD22124432
CAS číslo: 97776-06-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR