Methyl (S)-2-amino-3-(3-bromophenyl)propanoate hydrochloride 95%
Dodavatel: Apollo Scientific
OR471701-1G
OR471701-5G
APOSOR471701-1GEA
10350
CZK
APOSOR471701-1G
APOSOR471701-5G
Methyl (S)-2-amino-3-(3-bromophenyl)propanoate hydrochloride 95%
Methyl (S)-2-amino-3-(3-bromophenyl)propanoate hydrochloride
Vzorec:
C₁₀H₁₂BrNO₂·ClH MW: 294,573 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12911124 CAS číslo: 880347-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...