Tisk…

4-Amino-2-(tert-butyl)pyrimidine 95%

Dodavatel: Apollo Scientific

OR471697-1G OR471697-250MG
APOSOR471697-1GEA 20630 CZK
APOSOR471697-1G APOSOR471697-250MG
4-Amino-2-(tert-butyl)pyrimidine 95%
4-Amino-2-(tert-butyl)pyrimidine
Vzorec: C₈H₁₃N₃
MW: 151,209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646228
CAS číslo: 114362-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR