Tisk…

1,2,5-Thiadiazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR471696-1G OR471696-5G
APOSOR471696-1GEA 3060 CZK
APOSOR471696-1G APOSOR471696-5G
1,2,5-Thiadiazole-3-carboxylic acid 95%
1,2,5-Thiadiazole-3-carboxylic acid
Vzorec: C₃H₂N₂O₂S
MW: 130,125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01334979
CAS číslo: 13368-86-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR