Tisk…

3-Amino-3-(2-naphthyl)propionic acid 95%

Dodavatel: Apollo Scientific

Synonyms: DL-3-Amino-3-(2-naphthyl)propionic acid

OR471651-25G
APOSOR471651-25GEA 6620 CZK
APOSOR471651-25G
3-Amino-3-(2-naphthyl)propionic acid 95%
3-Amino-3-(2-naphthyl)propionic acid
Vzorec: C₁₃H₁₃NO₂
MW: 215,248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01871322
CAS číslo: 129042-57-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR