(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran 97%
Dodavatel: Apollo Scientific
Synonyms:
(S)-3-(4-(5-Bromo-2-chlorobenzyl)phenoxy)tetrahydrofuran
OR471641-25G
APOSOR471641-25GEA
4960
CZK
APOSOR471641-25G
(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran 97%
(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran
Vzorec:
C₁₇H₁₆BrClO₂ MW: 367,665 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27920793 CAS číslo: 915095-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...