3-Aminotetrahydrofuran-3-carbonitrile hydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR471596-1G
OR471596-5G
APOSOR471596-1GEA
4630
CZK
APOSOR471596-1G
APOSOR471596-5G
3-Aminotetrahydrofuran-3-carbonitrile hydrochloride 95%
3-Aminotetrahydrofuran-3-carbonitrile hydrochloride
Vzorec:
C₅H₈N₂O·ClH MW: 148,591 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20501945 CAS číslo: 1354963-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...