7-Bromo-5-methylindoline-2,3-dione
Dodavatel: Apollo Scientific
OR471591-1G
OR471591-5G
APOSOR471591-1GEA
4670
CZK
APOSOR471591-1G
APOSOR471591-5G
7-Bromo-5-methylindoline-2,3-dione
7-Bromo-5-methylindoline-2,3-dione
Vzorec:
C₉H₆BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00462954 CAS číslo: 108938-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...