Tisk…

N-Succinimidyl 3-Methoxy-4-nitrobenzoate

Dodavatel: Apollo Scientific

OR471578-1G OR471578-5G
APOSOR471578-1GEA 8700 CZK
APOSOR471578-1G APOSOR471578-5G
N-Succinimidyl 3-Methoxy-4-nitrobenzoate
N-Succinimidyl 3-Methoxy-4-nitrobenzoate
Vzorec: C₁₂H₁₀N₂O₇
MW: 294.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29065913
CAS číslo: 2006277-78-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR