3-(tert-Butyl)-2-hydroxy-5-(4-pyridyl)benzaldehyde
Dodavatel: Apollo Scientific
Synonyms:
3-(tert-Butyl)-2-hydroxy-5-(pyridin-4-yl)benzaldehyde
OR471573-1G
OR471573-250MG
OR471573-5G
APOSOR471573-1GEA
17130
CZK
APOSOR471573-1G
APOSOR471573-250MG
APOSOR471573-5G
3-(tert-Butyl)-2-hydroxy-5-(4-pyridyl)benzaldehyde
3-(tert-Butyl)-2-hydroxy-5-(4-pyridyl)benzaldehyde
Vzorec:
C₁₆H₁₇NO₂ MW: 255,312 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29065910 CAS číslo: 342037-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...