Tisk…

2-Chloro-3-iodopyrazine

Dodavatel: Apollo Scientific

OR471572-1G OR471572-5G
APOSOR471572-1GEA 5030 CZK
APOSOR471572-1G APOSOR471572-5G
2-Chloro-3-iodopyrazine
2-Chloro-3-iodopyrazine
Vzorec: C₄H₂ClIN₂
Storage Temperature: Ambient
MDL Number: MFCD09909706
CAS číslo: 191340-85-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR