6-Bromo-3-isopropyl-1H-indazole
Dodavatel: Apollo Scientific
OR471553-1G
OR471553-250MG
OR471553-5G
APOSOR471553-1GEA
26090
CZK
APOSOR471553-1G
APOSOR471553-250MG
APOSOR471553-5G
6-Bromo-3-isopropyl-1H-indazole
6-Bromo-3-isopropyl-1H-indazole
Vzorec:
C₁₀H₁₁BrN₂ MW: 239.112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27987866 CAS číslo: 1391123-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...