2,2-Dimethyl-4-oxochroman-7-carbonitrile
Dodavatel: Apollo Scientific
OR471548-1G
OR471548-250MG
OR471548-5G
APOSOR471548-1GEA
13920
CZK
APOSOR471548-1G
APOSOR471548-250MG
APOSOR471548-5G
2,2-Dimethyl-4-oxochroman-7-carbonitrile
2,2-Dimethyl-4-oxochroman-7-carbonitrile
Vzorec:
C₁₂H₁₁NO₂ MW: 201,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15527100 CAS číslo: 130200-02-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...