Tisk…
Methyl 4-(Nicotinamidomethyl)benzoate
Dodavatel: Apollo Scientific
OR471546-1G OR471546-5G
APOSOR471546-1GEA 10450 CZK
APOSOR471546-1G APOSOR471546-5G
Methyl 4-(Nicotinamidomethyl)benzoate
Methyl 4-(Nicotinamidomethyl)benzoate
Vzorec: C₁₅H₁₄N₂O₃
MW: 270.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03971539
CAS číslo: 554407-47-5