Tisk…

4-[[(4-Phenyl-1-piperazinyl)imino]methyl]-1,3-benzenediol

Dodavatel: Apollo Scientific

OR471542-1G OR471542-250MG OR471542-5G
APOSOR471542-1GEA 12770 CZK
APOSOR471542-1G APOSOR471542-250MG APOSOR471542-5G
4-[[(4-Phenyl-1-piperazinyl)imino]methyl]-1,3-benzenediol
4-[[(4-Phenyl-1-piperazinyl)imino]methyl]-1,3-benzenediol
Vzorec: C₁₇H₁₉N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01914767
CAS číslo: 306953-58-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR