3-[2-(Boc-amino)ethoxy]phenylboronic acid pinacol ester
Dodavatel: Apollo Scientific
OR471539-1G
OR471539-5G
OR471539-250MG
APOSOR471539-1GEA
10190
CZK
APOSOR471539-1G
APOSOR471539-5G
APOSOR471539-250MG
3-[2-(Boc-amino)ethoxy]phenylboronic acid pinacol ester
3-[2-(Boc-amino)ethoxy]phenylboronic acid pinacol ester
Vzorec:
C₁₉H₃₀BNO₅ MW: 363,256 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29055295 CAS číslo: 1505516-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...