3-(Cbz-aminomethyl)phenol
Dodavatel: Apollo Scientific
OR471537-25G
OR471537-5G
APOSOR471537-25GEA
28700
CZK
APOSOR471537-25G
APOSOR471537-5G
3-(Cbz-aminomethyl)phenol
3-(Cbz-aminomethyl)phenol
Vzorec:
C₁₅H₁₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD14585006 CAS číslo: 75383-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...