Methyl 3-(Boc-amino)pyrrolidine-3-carboxylate
Dodavatel: Apollo Scientific
Synonyms:
Methyl 3-((tert-butoxycarbonyl)amino)pyrrolidine-3-carboxylate
OR471536-1G
OR471536-250MG
APOSOR471536-1GEA
37690
CZK
APOSOR471536-1G
APOSOR471536-250MG
Methyl 3-(Boc-amino)pyrrolidine-3-carboxylate
Methyl 3-(Boc-amino)pyrrolidine-3-carboxylate
Vzorec:
C₁₁H₂₀N₂O₄ MW: 244,287 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26937551 CAS číslo: 1382035-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...