Tisk…

Ethyl 3-(Boc-amino)pyrrolidine-3-carboxylate

Dodavatel: Apollo Scientific

OR471535-1G OR471535-250MG
APOSOR471535-1GEA 37690 CZK
APOSOR471535-1G APOSOR471535-250MG
Ethyl 3-(Boc-amino)pyrrolidine-3-carboxylate
Ethyl 3-(Boc-amino)pyrrolidine-3-carboxylate
Vzorec: C₁₂H₂₂N₂O₄
MW: 258,314 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29055297
CAS číslo: 1613023-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR