Tisk…

5-Bromo-1-methylpyrazole-4-carbonitrile

Dodavatel: Apollo Scientific

OR471533-1G OR471533-250MG OR471533-5G
APOSOR471533-1GEA 3640 CZK
APOSOR471533-1G APOSOR471533-250MG APOSOR471533-5G
5-Bromo-1-methylpyrazole-4-carbonitrile
5-Bromo-1-methylpyrazole-4-carbonitrile
Vzorec: C₅H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD17168487
CAS číslo: 1269293-80-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR