Tisk…

2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide

Dodavatel: Apollo Scientific

OR471527-1G OR471527-250MG OR471527-5G
APOSOR471527-1GEA 19150 CZK
APOSOR471527-1G APOSOR471527-250MG APOSOR471527-5G
2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide
2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide
Vzorec: C₁₅H₁₃ClN₄O₅
Storage Temperature: Ambient
MDL Number: MFCD27918643
CAS číslo: 1079264-82-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR