Tisk…

4-(4-Hydroxybenzyl)tetrahydropyran-4-carboxylic acid

Dodavatel: Apollo Scientific

OR471525-1G OR471525-250MG OR471525-5G
APOSOR471525-1GEA 22920 CZK
APOSOR471525-1G APOSOR471525-250MG APOSOR471525-5G
4-(4-Hydroxybenzyl)tetrahydropyran-4-carboxylic acid
4-(4-Hydroxybenzyl)tetrahydropyran-4-carboxylic acid
Vzorec: C₁₃H₁₆O₄
MW: 236,264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23385393
CAS číslo: 1499681-44-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR