2-Chloro-5-(methoxymethoxy)pyridine
Dodavatel: Apollo Scientific
OR471503-1G
OR471503-250MG
OR471503-5G
APOSOR471503-1GEA
4140
CZK
APOSOR471503-1G
APOSOR471503-250MG
APOSOR471503-5G
2-Chloro-5-(methoxymethoxy)pyridine
2-Chloro-5-(methoxymethoxy)pyridine
Vzorec:
C₇H₈ClNO₂ MW: 173.597 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20278147 CAS číslo: 877133-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...