Tisk…

2-(Boc-amino)-1-methoxy-3-nitrobenzene

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl (2-methoxy-6-nitrophenyl)carbamate

OR471498-1G OR471498-250MG OR471498-5G
APOSOR471498-1GEA 3810 CZK
APOSOR471498-1G APOSOR471498-250MG APOSOR471498-5G
2-(Boc-amino)-1-methoxy-3-nitrobenzene
2-(Boc-amino)-1-methoxy-3-nitrobenzene
Vzorec: C₁₂H₁₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD29049234
CAS číslo: 1283176-56-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR