2-Chloro-6-methoxypurine
Dodavatel: Apollo Scientific
OR471479-1G
OR471479-250MG
APOSOR471479-1GEA
4470
CZK
APOSOR471479-1G
APOSOR471479-250MG
2-Chloro-6-methoxypurine
2-Chloro-6-methoxypurine
Vzorec:
C₆H₅ClN₄O MW: 184.583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20692004 CAS číslo: 1198-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...