Tisk…

6-Chloro-3-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one

Dodavatel: Apollo Scientific

OR471473-1G OR471473-250MG OR471473-5G
APOSOR471473-1GEA 20300 CZK
APOSOR471473-1G APOSOR471473-250MG APOSOR471473-5G
6-Chloro-3-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
6-Chloro-3-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
Vzorec: C₇H₆ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD01085385
CAS číslo: 370074-74-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR