Tisk…

4-Chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine hydrochloride

Dodavatel: Apollo Scientific

OR471463-1G OR471463-250MG
APOSOR471463-1GEA 26090 CZK
APOSOR471463-1G APOSOR471463-250MG
4-Chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine hydrochloride
4-Chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine hydrochloride
Vzorec: C₇H₈ClN₃·ClH
MW: 206,072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699257
CAS číslo: 2006277-76-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR